Learn More
Medchemexpress LLC Monomethyl auristatin E-d8 | 2070009-72-0 | MFCD28386277 | 99.3% | 726.03 | C39H59D8N5O7 | 10 MG
SDP
Catalog No. 5000287526
Encompass_Preferred
MMAE-d8 is a deuterated isotopologue of monomethyl auristatin E, a potent mitotic inhibitor that disrupts microtubule dynamics. The deuterium-labeled compound is designed for use as an analytical standard, tracer, or internal standard in antibody-drug conjugate research, mass spectrometry, and pharmacokinetic studies. Supplied as a high-purity solid for laboratory use.
  • Deuterated isotopologue for analytical and tracer applications.
  • Potent tubulin inhibitor useful as an ADC payload reference.
  • High reported purity suitable for sensitive assays.
  • Available in small milligram pack sizes for research workflows.
  • Supplied as a stable solid for accurate dosing and handling.
Catalog No. 50-002-87526 Supplier Medchemexpress LLC Supplier No. HY15162A10MG
MMAE-d8 is a deuterated isotopologue of monomethyl auristatin E, a potent mitotic inhibitor that disrupts microtubule dynamics. The deuterium-labeled compound is designed for use as an analytical standard, tracer, or internal standard in antibody-drug conjugate research, mass spectrometry, and pharmacokinetic studies. Supplied as a high-purity solid for laboratory use.
  • Deuterated isotopologue for analytical and tracer applications.
  • Potent tubulin inhibitor useful as an ADC payload reference.
  • High reported purity suitable for sensitive assays.
  • Available in small milligram pack sizes for research workflows.
  • Supplied as a stable solid for accurate dosing and handling.