Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-1 | 120205-48-3 | 99.5% | 295.85 | 1 ML
SDP
Catalog No. 5000404711
Encompass_Preferred
Monomethyl auristatin E intermediate-1 is an intermediate reagent used in the synthesis of Monomethyl auristatin E. This compound is a microtubule/tubulin inhibitor that exhibits anticancer activity and is widely utilized as the cytotoxic component in antibody-drug conjugates (ADCs).
  • Used as a cytotoxic component of antibody-drug conjugates (ADCs) to treat several different cancer types.
  • Microtubule/tubulin inhibitor with anticancer activity.
Catalog No. 50-004-04711 Supplier Medchemexpress LLC Supplier No. HY3304810MM/1ML
Monomethyl auristatin E intermediate-1 is an intermediate reagent used in the synthesis of Monomethyl auristatin E. This compound is a microtubule/tubulin inhibitor that exhibits anticancer activity and is widely utilized as the cytotoxic component in antibody-drug conjugates (ADCs).
  • Used as a cytotoxic component of antibody-drug conjugates (ADCs) to treat several different cancer types.
  • Microtubule/tubulin inhibitor with anticancer activity.