Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-1 | 120205-48-3 | 99.5% | 295.85 | 100 MG
SDP
Catalog No. 5000401559
Encompass_Preferred
Monomethyl auristatin E intermediate-1 is a key reagent for the synthesis of Monomethyl auristatin E (MMAE). MMAE is recognized as a potent microtubule/tubulin inhibitor with anticancer properties, commonly utilized as the cytotoxic component in antibody-drug conjugates (ADCs). This intermediate is provided as a solid, appearing white to off-white, and is suitable for research applications.
  • Intermediate reagent for Monomethyl auristatin E synthesis
  • MMAE is a microtubule/tubulin inhibitor
  • MMAE has anticancer properties
  • MMAE is used as a cytotoxic component in antibody-drug conjugates
  • Appears as a white to off-white solid
  • For research use only
Catalog No. 50-004-01559 Supplier Medchemexpress LLC Supplier No. HY33048100MG
Monomethyl auristatin E intermediate-1 is a key reagent for the synthesis of Monomethyl auristatin E (MMAE). MMAE is recognized as a potent microtubule/tubulin inhibitor with anticancer properties, commonly utilized as the cytotoxic component in antibody-drug conjugates (ADCs). This intermediate is provided as a solid, appearing white to off-white, and is suitable for research applications.
  • Intermediate reagent for Monomethyl auristatin E synthesis
  • MMAE is a microtubule/tubulin inhibitor
  • MMAE has anticancer properties
  • MMAE is used as a cytotoxic component in antibody-drug conjugates
  • Appears as a white to off-white solid
  • For research use only