Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-1 | 120205-48-3 | 99.5% | 295.85 | 25 G
SDP
Catalog No. 5000403069
Encompass_Preferred
Monomethyl auristatin E intermediate-1 is an intermediate reagent used in the synthesis of Monomethyl auristatin E. Monomethyl auristatin E (MMAE) is a microtubule/tubulin inhibitor with anticancer activity and is widely utilized as the cytotoxic component (ADC Cytotoxin) of antibody-drug conjugates (ADCs).
  • Intermediate reagent in the synthesis of Monomethyl auristatin E.
  • Inhibits microtubules and tubulin.
  • Exhibits anticancer activity.
  • Used as the cytotoxic component of antibody-drug conjugates.
Catalog No. 50-004-03069 Supplier Medchemexpress LLC Supplier No. 25GHY33048
Monomethyl auristatin E intermediate-1 is an intermediate reagent used in the synthesis of Monomethyl auristatin E. Monomethyl auristatin E (MMAE) is a microtubule/tubulin inhibitor with anticancer activity and is widely utilized as the cytotoxic component (ADC Cytotoxin) of antibody-drug conjugates (ADCs).
  • Intermediate reagent in the synthesis of Monomethyl auristatin E.
  • Inhibits microtubules and tubulin.
  • Exhibits anticancer activity.
  • Used as the cytotoxic component of antibody-drug conjugates.