Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethyl auristatin E intermediate-1 | 120205-48-3 | 99.5% | 295.85 | 5 G
SDP

Catalog No. 5000401485
Encompass_Preferred

Monomethyl auristatin E intermediate-1 is an intermediate reagent used in the synthesis of Monomethyl auristatin E (MMAE). MMAE is a microtubule/tubulin inhibitor with anticancer activity, and it is widely utilized as the cytotoxic component (ADC Cytotoxin) of antibody-drug conjugates (ADCs). This product is for research use only and not for sale to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C14H30ClNO3
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, stored under nitrogen, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen, away from moisture).

Catalog No. 50-004-01485 Supplier Medchemexpress LLC Supplier No. HY330485G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monomethyl auristatin E intermediate-1 is an intermediate reagent used in the synthesis of Monomethyl auristatin E (MMAE). MMAE is a microtubule/tubulin inhibitor with anticancer activity, and it is widely utilized as the cytotoxic component (ADC Cytotoxin) of antibody-drug conjugates (ADCs). This product is for research use only and not for sale to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C14H30ClNO3
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, stored under nitrogen, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen, away from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.