Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-1 | 120205-48-3 | HY-33048 | 99.5% | 295.85 | 250 MG
SDP
Catalog No. 5000402158
Encompass_Preferred
Monomethyl auristatin E intermediate-1 is an intermediate reagent in the synthesis of Monomethyl auristatin E. Monomethyl auristatin E (MMAE) is a microtubule/tubulin inhibitor with anticancer activity and is widely used as the cytotoxic component of antibody-drug conjugates (ADCs) to treat several different cancer types.
  • Intermediate reagent in synthesis
  • Inhibits microtubule/tubulin
  • Exhibits anticancer activity
  • Used as a cytotoxic component of antibody-drug conjugates
Catalog No. 50-004-02158 Supplier Medchemexpress LLC Supplier No. 250MGHY33048
Monomethyl auristatin E intermediate-1 is an intermediate reagent in the synthesis of Monomethyl auristatin E. Monomethyl auristatin E (MMAE) is a microtubule/tubulin inhibitor with anticancer activity and is widely used as the cytotoxic component of antibody-drug conjugates (ADCs) to treat several different cancer types.
  • Intermediate reagent in synthesis
  • Inhibits microtubule/tubulin
  • Exhibits anticancer activity
  • Used as a cytotoxic component of antibody-drug conjugates