Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-11 | 870640-62-3 | 98.8% | C14H23NO | 250 MG
SDP
Catalog No. 5000358390
Encompass_Preferred
Monomethyl auristatin E intermediate-11 is an intermediate reactant used in the synthesis of Monomethyl auristatin E (HY-15162). Monomethyl auristatin E (MMAE) acts as a microtubule/tubulin inhibitor and exhibits anticancer activity. MMAE is commonly utilized as the cytotoxic component (ADC Cytotoxin) in antibody-drug conjugates (ADCs).
  • Intermediate reactant in Monomethyl auristatin E synthesis
  • Monomethyl auristatin E (MMAE) acts as a microtubule/tubulin inhibitor
  • Exhibits anticancer activity
  • Utilized as cytotoxic component in antibody-drug conjugates (ADCs)
Catalog No. 50-003-58390 Supplier Medchemexpress LLC Supplier No. 250MGHY78908
Monomethyl auristatin E intermediate-11 is an intermediate reactant used in the synthesis of Monomethyl auristatin E (HY-15162). Monomethyl auristatin E (MMAE) acts as a microtubule/tubulin inhibitor and exhibits anticancer activity. MMAE is commonly utilized as the cytotoxic component (ADC Cytotoxin) in antibody-drug conjugates (ADCs).
  • Intermediate reactant in Monomethyl auristatin E synthesis
  • Monomethyl auristatin E (MMAE) acts as a microtubule/tubulin inhibitor
  • Exhibits anticancer activity
  • Utilized as cytotoxic component in antibody-drug conjugates (ADCs)