Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethyl auristatin E intermediate-11 | 870640-62-3 | 98.8% | C14H23NO | 50 MG
SDP

Catalog No. 5000435031
Encompass_Preferred

Monomethyl auristatin E intermediate-11 is an intermediate reactant in the synthesis of Monomethyl auristatin E. Monomethyl auristatin E is a microtubule/tubulin inhibitor with anticancer activity and is widely used as the cytotoxic component (ADC Cytotoxin) of antibody-drug conjugates (ADCs).

  • Intermediate for Monomethyl auristatin E synthesis
  • Microtubule/tubulin inhibitor
  • Used as cytotoxic component in antibody-drug conjugates (ADCs)
  • Appears as colorless to light yellow oil
  • Molecular weight: 221.34
  • Purity: 98.8%
  • Store pure form at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-35031 Supplier Medchemexpress LLC Supplier No. HY7890850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monomethyl auristatin E intermediate-11 is an intermediate reactant in the synthesis of Monomethyl auristatin E. Monomethyl auristatin E is a microtubule/tubulin inhibitor with anticancer activity and is widely used as the cytotoxic component (ADC Cytotoxin) of antibody-drug conjugates (ADCs).

  • Intermediate for Monomethyl auristatin E synthesis
  • Microtubule/tubulin inhibitor
  • Used as cytotoxic component in antibody-drug conjugates (ADCs)
  • Appears as colorless to light yellow oil
  • Molecular weight: 221.34
  • Purity: 98.8%
  • Store pure form at -20°C for 3 years, 4°C for 2 years
  • Store in solvent at -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.