Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-14 | 1932791-56-4 | >98.5% | C13H19NO2 | 25 MG
SDP
Catalog No. 5000358878
Encompass_Preferred
Monomethyl auristatin E intermediate-14 is an intermediate reagent used in the synthesis of Monomethyl auristatin E (MMAE). MMAE is a microtubule/tubulin inhibitor with anticancer activity and serves as the cytotoxic component of antibody-drug conjugates (ADCs).
  • Intermediate reagent for Monomethyl auristatin E synthesis
  • Microtubule/tubulin inhibitor
  • Exhibits anticancer activity
  • Cytotoxic component of antibody-drug conjugates (ADCs)
Catalog No. 50-003-58878 Supplier Medchemexpress LLC Supplier No. 25MGHY78913
Monomethyl auristatin E intermediate-14 is an intermediate reagent used in the synthesis of Monomethyl auristatin E (MMAE). MMAE is a microtubule/tubulin inhibitor with anticancer activity and serves as the cytotoxic component of antibody-drug conjugates (ADCs).
  • Intermediate reagent for Monomethyl auristatin E synthesis
  • Microtubule/tubulin inhibitor
  • Exhibits anticancer activity
  • Cytotoxic component of antibody-drug conjugates (ADCs)