Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-14 | 1932791-56-4 | C13H19NO2 | 50 MG
SDP
Catalog No. 5000433002
Encompass_Preferred
Monomethyl auristatin E intermediate-14 is a crucial reagent utilized in the synthesis of Monomethyl auristatin E. This intermediate supports the production of MMAE, a potent microtubule/tubulin inhibitor recognized for its significant anticancer activity. MMAE is frequently incorporated as the cytotoxic component in antibody-drug conjugates (ADCs).
  • Intermediate for Monomethyl auristatin E synthesis
  • Supports production of a microtubule/tubulin inhibitor
  • Aids in the creation of anticancer agents
  • Can be used as a component in antibody-drug conjugates
Catalog No. 50-004-33002 Supplier Medchemexpress LLC Supplier No. HY7891350MG
Monomethyl auristatin E intermediate-14 is a crucial reagent utilized in the synthesis of Monomethyl auristatin E. This intermediate supports the production of MMAE, a potent microtubule/tubulin inhibitor recognized for its significant anticancer activity. MMAE is frequently incorporated as the cytotoxic component in antibody-drug conjugates (ADCs).
  • Intermediate for Monomethyl auristatin E synthesis
  • Supports production of a microtubule/tubulin inhibitor
  • Aids in the creation of anticancer agents
  • Can be used as a component in antibody-drug conjugates