Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethyl auristatin E intermediate-15 | 173653-47-9 | C20H29NO5 | 50 MG
SDP

Catalog No. 5000433058
Encompass_Preferred

This product is an oil identified for use as laboratory chemicals and in the manufacture of substances. It should be stored in a tightly sealed container in a cool, well-ventilated area, away from direct sunlight and sources of ignition. It is stable under recommended storage conditions.

  • Oil appearance
  • Molecular weight: 363.45
  • For laboratory chemical use
  • For substance manufacturing
  • Store at -20°C for 3 years or 4°C for 2 years (pure form)
  • Store at -80°C for 6 months or -20°C for 1 month (in solvent)
  • Shipping at room temperature for less than 2 weeks
  • Causes skin and serious eye irritation
  • May cause respiratory irritation
  • Harmful if swallowed

Catalog No. 50-004-33058 Supplier Medchemexpress LLC Supplier No. HY7919650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This product is an oil identified for use as laboratory chemicals and in the manufacture of substances. It should be stored in a tightly sealed container in a cool, well-ventilated area, away from direct sunlight and sources of ignition. It is stable under recommended storage conditions.

  • Oil appearance
  • Molecular weight: 363.45
  • For laboratory chemical use
  • For substance manufacturing
  • Store at -20°C for 3 years or 4°C for 2 years (pure form)
  • Store at -80°C for 6 months or -20°C for 1 month (in solvent)
  • Shipping at room temperature for less than 2 weeks
  • Causes skin and serious eye irritation
  • May cause respiratory irritation
  • Harmful if swallowed

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.