Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethyl auristatin E intermediate-2 | 317.38 | C15H27NO6 | 100 MG
SDP

Catalog No. 5000202346
Encompass_Preferred

Monomethyl auristatin E intermediate-2 is an intermediate reagent used in the synthesis of monomethyl auristatin E, a potent antimitotic agent commonly used as an antibody-drug conjugate (ADC) payload. Supplied as a 100 mg solid-liquid mixture; consult the product datasheet and SDS for batch-specific purity and handling instructions.

  • Used as a synthetic intermediate for monomethyl auristatin E
  • Provided in a 100 mg package
  • Molecular weight: 317.38
  • Chemical formula: C15H27NO6
  • Appearance: white to off-white solid-liquid mixture
  • Refer to the datasheet and SDS for purity and handling instructions

Catalog No. 50-002-02346 Supplier Medchemexpress LLC Supplier No. HY49413100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monomethyl auristatin E intermediate-2 is an intermediate reagent used in the synthesis of monomethyl auristatin E, a potent antimitotic agent commonly used as an antibody-drug conjugate (ADC) payload. Supplied as a 100 mg solid-liquid mixture; consult the product datasheet and SDS for batch-specific purity and handling instructions.

  • Used as a synthetic intermediate for monomethyl auristatin E
  • Provided in a 100 mg package
  • Molecular weight: 317.38
  • Chemical formula: C15H27NO6
  • Appearance: white to off-white solid-liquid mixture
  • Refer to the datasheet and SDS for purity and handling instructions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.