Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-3 | 265.30 | 100 MG
SDP
Catalog No. 5000404379
Encompass_Preferred
Monomethyl auristatin E intermediate-3 is an intermediate reagent in the synthesis of Monomethyl auristatin E (HY-15162). Monomethyl auristatin E (MMAE) is a microtubule/tubulin inhibitor with anticancer activity. MMAE is widely used as the cytotoxic component (ADC Cytotoxin) of antibody-drug conjugates (ADCs).
  • Appearance: Oil
  • Color: Colorless to light yellow
Catalog No. 50-004-04379 Supplier Medchemexpress LLC Supplier No. HY43146100MG
Monomethyl auristatin E intermediate-3 is an intermediate reagent in the synthesis of Monomethyl auristatin E (HY-15162). Monomethyl auristatin E (MMAE) is a microtubule/tubulin inhibitor with anticancer activity. MMAE is widely used as the cytotoxic component (ADC Cytotoxin) of antibody-drug conjugates (ADCs).
  • Appearance: Oil
  • Color: Colorless to light yellow