Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-3 | 265.30 | 250 MG
SDP
Catalog No. 5000402982
Encompass_Preferred
Monomethyl auristatin E intermediate-3 is a reagent used in the synthesis of Monomethyl auristatin E (MMAE). MMAE acts as a microtubule/tubulin inhibitor, demonstrating anticancer activity and is commonly employed as the cytotoxic component in antibody-drug conjugates (ADCs).
  • Intermediate reagent for Monomethyl auristatin E synthesis.
  • Precursor to a microtubule/tubulin inhibitor.
  • Used in antibody-drug conjugates (ADCs) as a cytotoxic component.
Catalog No. 50-004-02982 Supplier Medchemexpress LLC Supplier No. 250MGHY43146
Monomethyl auristatin E intermediate-3 is a reagent used in the synthesis of Monomethyl auristatin E (MMAE). MMAE acts as a microtubule/tubulin inhibitor, demonstrating anticancer activity and is commonly employed as the cytotoxic component in antibody-drug conjugates (ADCs).
  • Intermediate reagent for Monomethyl auristatin E synthesis.
  • Precursor to a microtubule/tubulin inhibitor.
  • Used in antibody-drug conjugates (ADCs) as a cytotoxic component.