Learn More
Medchemexpress LLC Monomethyl auristatin E intermediate-9 | 120205-58-5 | >98.0% | 393.52 | C22H35NO5 | 10 MG
SDP
Catalog No. 5000298501
Encompass_Preferred
Monomethyl auristatin E intermediate-9 is a synthetic chemical intermediate supplied as a dry powder for laboratory research. It is used in the preparation of monomethyl auristatin E (MMAE), a potent microtubule/tubulin inhibitor commonly employed as the cytotoxic component of antibody-drug conjugates (ADCs).
  • Serves as an intermediate for synthesizing monomethyl auristatin E (MMAE).
  • Intended for research use only; not for human or veterinary use.
  • Suitable for ADC cytotoxin development and tubulin inhibition studies.
  • Molecular formula C22H35NO5; molecular weight 393.52.
  • Typical purity >98.0%.
  • Available in small research pack sizes, e.g., 5 MG and 10 MG.
Catalog No. 50-002-98501 Supplier Medchemexpress LLC Supplier No. HY7890310MG
Monomethyl auristatin E intermediate-9 is a synthetic chemical intermediate supplied as a dry powder for laboratory research. It is used in the preparation of monomethyl auristatin E (MMAE), a potent microtubule/tubulin inhibitor commonly employed as the cytotoxic component of antibody-drug conjugates (ADCs).
  • Serves as an intermediate for synthesizing monomethyl auristatin E (MMAE).
  • Intended for research use only; not for human or veterinary use.
  • Suitable for ADC cytotoxin development and tubulin inhibition studies.
  • Molecular formula C22H35NO5; molecular weight 393.52.
  • Typical purity >98.0%.
  • Available in small research pack sizes, e.g., 5 MG and 10 MG.