Learn More
Medchemexpress LLC Monomethyl auristatin E (MMAE) | 474645-27-7 | 100.0% | 250 MG
SDP
Catalog No. 5000358058
Encompass_Preferred
Monomethyl auristatin E (MMAE), also known as SGD-1010, is a synthetic derivative of dolastatin 10. It acts as a potent mitotic inhibitor by preventing tubulin polymerization. This compound is extensively used as a cytotoxic component in antibody-drug conjugates (ADCs) for the treatment of various cancer types.
  • Potent mitotic inhibitor.
  • Inhibits tubulin polymerization.
  • Widely used as a cytotoxic component in antibody-drug conjugates (ADCs).
  • Effective in treating several different cancer types.
Catalog No. 50-003-58058 Supplier Medchemexpress LLC Supplier No. 250MGHY78901
Add to Cart
Add to Cart
Monomethyl auristatin E (MMAE), also known as SGD-1010, is a synthetic derivative of dolastatin 10. It acts as a potent mitotic inhibitor by preventing tubulin polymerization. This compound is extensively used as a cytotoxic component in antibody-drug conjugates (ADCs) for the treatment of various cancer types.
  • Potent mitotic inhibitor.
  • Inhibits tubulin polymerization.
  • Widely used as a cytotoxic component in antibody-drug conjugates (ADCs).
  • Effective in treating several different cancer types.