Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethyl auristatin E-SMCC conjugate | 2021179-11-1 | 95.7% | 1,140.43 | C58H89N7O14S | 10 MG
SDP

Catalog No. 5000203794
Encompass_Preferred

MMAE-SMCC is a drug-linker conjugate used to prepare antibody-drug conjugates (ADCs). It links monomethyl auristatin E (a potent antimitotic tubulin inhibitor) to the SMCC linker to enable covalent attachment to antibodies for targeted cytotoxic delivery.

  • White to off-white solid.
  • Purity 95.7%.
  • Molecular weight 1,140.43 and formula C58H89N7O14S.
  • CAS number 2021179-11-1.
  • Solubility DMSO: 100 mg/mL (approx. 87.69 mM); ultrasonic assistance recommended.
  • Storage -20 °C; protect from light; store under nitrogen.
  • Unstable in solution; freshly prepare working solutions.

Catalog No. 50-002-03794 Supplier Medchemexpress LLC Supplier No. HY13566010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MMAE-SMCC is a drug-linker conjugate used to prepare antibody-drug conjugates (ADCs). It links monomethyl auristatin E (a potent antimitotic tubulin inhibitor) to the SMCC linker to enable covalent attachment to antibodies for targeted cytotoxic delivery.

  • White to off-white solid.
  • Purity 95.7%.
  • Molecular weight 1,140.43 and formula C58H89N7O14S.
  • CAS number 2021179-11-1.
  • Solubility DMSO: 100 mg/mL (approx. 87.69 mM); ultrasonic assistance recommended.
  • Storage -20 °C; protect from light; store under nitrogen.
  • Unstable in solution; freshly prepare working solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.