Learn More
Selleck Chemical LLC Monomethyl Fumarate
SDP
Supplier:  Selleck Chemical LLC S6889200MG
Encompass_Preferred
Monomethyl Fumarate (MMF Monomethylfumarate Fumaric acid monomethyl ester Methyl hydrogen fumarate) the active metabolite of the psoriasis drug Fumaderm is a potent GPR109A agonist Monomethyl Fumarate prevents major dysfunctions associated with neurodegenerative diseases oxidative stress mitochondrial dysfunction apoptosis and autophagy
Catalog No. 50-000-02849