Learn More
Medchemexpress LLC Monomethyl fumarate | 1616345-41-5 | MFCD00063174 | 100.0% | 130.10 g/mol | C5H6O4 | 5 MG
SDP
Catalog No. 5000247051
Encompass_Preferred
Monomethyl fumarate is the methyl ester metabolite of dimethyl fumarate and is used as a research compound and GPR109A agonist. The material is supplied for laboratory research and analytical reference; available as solid and as DMSO solutions, with analytical documentation available (COA/SDS).
  • High purity (≈99.95%).
  • Molecular formula C5H6O4 and molecular weight 130.10 g/mol.
  • Available as solid, free sample, and 10 mM solutions in DMSO.
  • Suitable as a reference standard and for in vitro pharmacology assays.
  • Not for human therapeutic or clinical use; research use only.
Catalog No. 50-002-47051 Supplier Medchemexpress LLC Supplier No. HY103252S5MG
Monomethyl fumarate is the methyl ester metabolite of dimethyl fumarate and is used as a research compound and GPR109A agonist. The material is supplied for laboratory research and analytical reference; available as solid and as DMSO solutions, with analytical documentation available (COA/SDS).
  • High purity (≈99.95%).
  • Molecular formula C5H6O4 and molecular weight 130.10 g/mol.
  • Available as solid, free sample, and 10 mM solutions in DMSO.
  • Suitable as a reference standard and for in vitro pharmacology assays.
  • Not for human therapeutic or clinical use; research use only.