Learn More
Medchemexpress LLC Monomethyl fumarate | 2756-87-8 | MFCD00063174 | 100.0% | 130.10 g/mol | C5H6O4 | 1 ML
SDP
Catalog No. 5000247083
Encompass_Preferred
Monomethyl fumarate is the methyl ester of fumaric acid and the active metabolite of dimethyl fumarate; it acts as a GPR109A agonist and is used in biochemical and pharmacological research. The product is offered with high purity and is available as a ready-to-use solution or as solid material for analytical and preparative applications.
  • High purity suitable for research applications.
  • Provided as a ready-to-use 10 mM solution in DMSO (1 mL).
  • Also available in solid form for bulk use or formulation.
  • Well-characterized molecular data: chemical formula C5H6O4 and molecular weight 130.10 g/mol.
  • Appropriate for use as a reference compound and in studies of GPR109A-mediated pathways.
Catalog No. 50-002-47083 Supplier Medchemexpress LLC Supplier No. HY10325210MM 1ML
Monomethyl fumarate is the methyl ester of fumaric acid and the active metabolite of dimethyl fumarate; it acts as a GPR109A agonist and is used in biochemical and pharmacological research. The product is offered with high purity and is available as a ready-to-use solution or as solid material for analytical and preparative applications.
  • High purity suitable for research applications.
  • Provided as a ready-to-use 10 mM solution in DMSO (1 mL).
  • Also available in solid form for bulk use or formulation.
  • Well-characterized molecular data: chemical formula C5H6O4 and molecular weight 130.10 g/mol.
  • Appropriate for use as a reference compound and in studies of GPR109A-mediated pathways.