Learn More
Medchemexpress LLC Monomethyl fumarate-d2 | 253122-44-0 | MFCD00063174 | 98.2% | 132.11 | C5H4D2O4 | 1 MG
SDP
Catalog No. 5000247179
Encompass_Preferred
Monomethyl fumarate-d2 is the deuterium-labeled form of monomethyl fumarate, provided as a high-purity research chemical and isotopically labeled standard for metabolic, pharmacological, and analytical studies.
  • Deuterium-labeled analog for isotope tracing and mass spectrometry.
  • High purity suitable for research applications.
  • Supplied in milligram quantities for small-scale experiments.
  • Useful as a metabolic standard and mechanistic probe.
Catalog No. 50-002-47179 Supplier Medchemexpress LLC Supplier No. HY103252S21MG
Monomethyl fumarate-d2 is the deuterium-labeled form of monomethyl fumarate, provided as a high-purity research chemical and isotopically labeled standard for metabolic, pharmacological, and analytical studies.
  • Deuterium-labeled analog for isotope tracing and mass spectrometry.
  • High purity suitable for research applications.
  • Supplied in milligram quantities for small-scale experiments.
  • Useful as a metabolic standard and mechanistic probe.