Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethyl fumarate-d3 | 1616345-41-5 | MFCD34469902 | 99.0% | 133.117 g/mol | C5H3D3O4 | 1 MG
SDP

Catalog No. 5000247041
Encompass_Preferred

Monomethyl fumarate-d3 is the deuterium-labeled analogue of monomethyl fumarate, supplied as an isotopically labeled tracer and internal standard for analytical and pharmacokinetic studies. It is intended for research use in quantitative workflows where precise isotope labeling improves detection and quantitation.

  • Isotopically labeled internal standard for NMR, GC-MS, and LC-MS.
  • Useful as a tracer in pharmacokinetic and metabolic profiling.
  • High analytical purity (≈99.0%).
  • Provided in small research quantities suitable for analytical assays.
  • Molecular formula C5H3D3O4; CAS 1616345-41-5.

Catalog No. 50-002-47041 Supplier Medchemexpress LLC Supplier No. HY103252S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monomethyl fumarate-d3 is the deuterium-labeled analogue of monomethyl fumarate, supplied as an isotopically labeled tracer and internal standard for analytical and pharmacokinetic studies. It is intended for research use in quantitative workflows where precise isotope labeling improves detection and quantitation.

  • Isotopically labeled internal standard for NMR, GC-MS, and LC-MS.
  • Useful as a tracer in pharmacokinetic and metabolic profiling.
  • High analytical purity (≈99.0%).
  • Provided in small research quantities suitable for analytical assays.
  • Molecular formula C5H3D3O4; CAS 1616345-41-5.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.