Learn More
ABclonal Technology MonoMethyl-Histone H3-K9
SDP
Catalog No. 5000006884
Encompass_Preferred
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A H2B H3 and H4) The chromatin fiber is further compacted through the interaction of a linker histone H1 with the DNA between the nucleosomes to form higher order chromatin structures This gene is intronless and encodes a replication-dependent histone that is a member of the histone H3 family Transcripts from this gene lack polyA tails instead they contain a palindromic termination element This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21 3
Catalog No. 50-000-06884 Supplier ABclonal Technology Supplier No. A22079100UL
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A H2B H3 and H4) The chromatin fiber is further compacted through the interaction of a linker histone H1 with the DNA between the nucleosomes to form higher order chromatin structures This gene is intronless and encodes a replication-dependent histone that is a member of the histone H3 family Transcripts from this gene lack polyA tails instead they contain a palindromic termination element This gene is located separately from the other H3 genes that are in the histone gene cluster on chromosome 6p22-p21 3