Learn More
ABclonal Technology MonoMethyl-Histone H4-K20
SDP
Catalog No. 5000007760
Encompass_Preferred
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes This structure consists of approximately 146 bp of DNA wrapped around a nucleosome an octamer composed of pairs of each of the four core histones (H2A H2B H3 and H4) The chromatin fiber is further compacted through the interaction of a linker histone H1 with the DNA between the nucleosomes to form higher order chromatin structures This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family Transcripts from this gene lack polyA tails instead they contain a palindromic termination element This gene is found in a histone cluster on chromosome 1 This gene is one of four histone genes in the cluster that are duplicated this record represents the centromeric copy
Catalog No. 50-000-07760 Supplier ABclonal Technology Supplier No. A22572100UL
Add to Cart
Add to Cart
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes This structure consists of approximately 146 bp of DNA wrapped around a nucleosome an octamer composed of pairs of each of the four core histones (H2A H2B H3 and H4) The chromatin fiber is further compacted through the interaction of a linker histone H1 with the DNA between the nucleosomes to form higher order chromatin structures This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family Transcripts from this gene lack polyA tails instead they contain a palindromic termination element This gene is found in a histone cluster on chromosome 1 This gene is one of four histone genes in the cluster that are duplicated this record represents the centromeric copy