Learn More
Medchemexpress LLC Monomethyl itaconate 10g
SDP
Catalog No. 5000206580
Encompass_Preferred
Monomethyl itaconate ( -Methyl itaconate) is a monomethyl ester derivative of Itaconic acid (HY-Y0520) Monomethyl itaconate can copolymerize with PEGMA to form pH-sensitive hydrogels[1]
Catalog No. 50-002-06580 Supplier Medchemexpress LLC Supplier No. HYW09421410G
Monomethyl itaconate ( -Methyl itaconate) is a monomethyl ester derivative of Itaconic acid (HY-Y0520) Monomethyl itaconate can copolymerize with PEGMA to form pH-sensitive hydrogels[1]