Learn More
Medchemexpress LLC Monomethyl phthalate | 4376-18-5 | MFCD00002466 | 180.16 | C9H8O4 | 100mg
SDP
Catalog No. 5000268004
Encompass_Preferred
Monomethyl phthalate is an orally active metabolite of phthalic acid Monomethyl phthalate as a urine biomarker after exposure to phthalate can be used as a detection indicator for thyroid cancer and benign nodules Monomethyl phthalate reduces the survival rate of young frogs Monomethyl phthalate induces oxidative damage to red blood cells in rats[1][2][3]
Catalog No. 50-002-68004 Supplier Medchemexpress LLC Supplier No. HYY1097100MG
Monomethyl phthalate is an orally active metabolite of phthalic acid Monomethyl phthalate as a urine biomarker after exposure to phthalate can be used as a detection indicator for thyroid cancer and benign nodules Monomethyl phthalate reduces the survival rate of young frogs Monomethyl phthalate induces oxidative damage to red blood cells in rats[1][2][3]