Learn More
Medchemexpress LLC Monomethyl phthalate | 4376-18-5 | MFCD00002466 | 180.16 | C9H8O4 | 1mL
SDP
Catalog No. 5000268082
Encompass_Preferred
Monomethyl phthalate is an orally active metabolite of phthalic acid Monomethyl phthalate as a urine biomarker after exposure to phthalate can be used as a detection indicator for thyroid cancer and benign nodules Monomethyl phthalate reduces the survival rate of young frogs Monomethyl phthalate induces oxidative damage to red blood cells in rats[1][2][3]
Catalog No. 50-002-68082 Supplier Medchemexpress LLC Supplier No. HYY109710MM/1ML
Monomethyl phthalate is an orally active metabolite of phthalic acid Monomethyl phthalate as a urine biomarker after exposure to phthalate can be used as a detection indicator for thyroid cancer and benign nodules Monomethyl phthalate reduces the survival rate of young frogs Monomethyl phthalate induces oxidative damage to red blood cells in rats[1][2][3]