Learn More
Medchemexpress LLC Monomethyl phthalate-d4 | 1276197-40-0 | 184.18 g/mol | C9H4D4O4 | 1mg
SDP
Catalog No. 5000267675
Encompass_Preferred
Monomethyl phthalate-d4 (2-(Methoxycarbonyl)benzoic acid-d4) is the deuterium labeled Monomethyl phthalate Monomethyl phthalate is a phthalate metabolite Monomethyl phthalate acts as a urinary biomarker of phthalates exposure and can be used as a standard for the determination of thyroid cancer and benign nodule[1]
Catalog No. 50-002-67675 Supplier Medchemexpress LLC Supplier No. HYY1097S1MG
Monomethyl phthalate-d4 (2-(Methoxycarbonyl)benzoic acid-d4) is the deuterium labeled Monomethyl phthalate Monomethyl phthalate is a phthalate metabolite Monomethyl phthalate acts as a urinary biomarker of phthalates exposure and can be used as a standard for the determination of thyroid cancer and benign nodule[1]