Learn More
Medchemexpress LLC Monomethyl phthalate-d4 | 1276197-40-0 | 97.6% | 184.18 | C9H4D4O4 | 10mg
SDP
Catalog No. 5000267095
Encompass_Preferred
Monomethyl phthalate-d4 (2-(Methoxycarbonyl)benzoic acid-d4) is the deuterium labeled Monomethyl phthalate Monomethyl phthalate is a phthalate metabolite Monomethyl phthalate acts as a urinary biomarker of phthalates exposure and can be used as a standard for the determination of thyroid cancer and benign nodule[1]
Catalog No. 50-002-67095 Supplier Medchemexpress LLC Supplier No. HYY1097S10MG
Monomethyl phthalate-d4 (2-(Methoxycarbonyl)benzoic acid-d4) is the deuterium labeled Monomethyl phthalate Monomethyl phthalate is a phthalate metabolite Monomethyl phthalate acts as a urinary biomarker of phthalates exposure and can be used as a standard for the determination of thyroid cancer and benign nodule[1]