Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethylauristatin F | 745017-94-1 | MFCD25976742 | >95.0% | 731.96 g/mol | C39H65N5O8 | 50 MG
SDP

Catalog No. 5000204344
Encompass_Preferred

MMAF (Monomethylauristatin F) is a synthetic antimitotic agent that inhibits tubulin polymerization and is widely used as a cytotoxic payload in antibody-drug conjugates (ADCs) and for antitumor research. It is supplied for research use and should be handled under appropriate safety precautions.

  • Potent tubulin polymerization inhibitor.
  • Used as a cytotoxic payload in antibody-drug conjugates (ADCs).
  • Cas number 745017-94-1.
  • Molecular formula C39H65N5O8; molecular weight 731.96 g/mol.
  • Typical storage: 4°C, sealed and protected from moisture.
  • Common research pack size: 50 mg.

Catalog No. 50-002-04344 Supplier Medchemexpress LLC Supplier No. HY1557950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MMAF (Monomethylauristatin F) is a synthetic antimitotic agent that inhibits tubulin polymerization and is widely used as a cytotoxic payload in antibody-drug conjugates (ADCs) and for antitumor research. It is supplied for research use and should be handled under appropriate safety precautions.

  • Potent tubulin polymerization inhibitor.
  • Used as a cytotoxic payload in antibody-drug conjugates (ADCs).
  • Cas number 745017-94-1.
  • Molecular formula C39H65N5O8; molecular weight 731.96 g/mol.
  • Typical storage: 4°C, sealed and protected from moisture.
  • Common research pack size: 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.