Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethylauristatin F hydrochloride | 1415246-68-2 | 768.42 | C39H66ClN5O8 | 50 MG
SDP

Catalog No. 5000201874
Encompass_Preferred

MMAF hydrochloride (Monomethylauristatin F hydrochloride) is a potent tubulin polymerization inhibitor used as an antitumor agent and as the cytotoxic payload in antibody-drug conjugates (ADCs). It is supplied in small milligram-scale amounts for research and ADC development.

  • Potent tubulin polymerization inhibitor.
  • Used as a cytotoxic component in ADCs.
  • CAS number 1415246-68-2.
  • Molecular weight 768.42.
  • Chemical formula C39H66ClN5O8.
  • Supplied as a solid in milligram quantities for research use.

Catalog No. 50-002-01874 Supplier Medchemexpress LLC Supplier No. HY15579A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MMAF hydrochloride (Monomethylauristatin F hydrochloride) is a potent tubulin polymerization inhibitor used as an antitumor agent and as the cytotoxic payload in antibody-drug conjugates (ADCs). It is supplied in small milligram-scale amounts for research and ADC development.

  • Potent tubulin polymerization inhibitor.
  • Used as a cytotoxic component in ADCs.
  • CAS number 1415246-68-2.
  • Molecular weight 768.42.
  • Chemical formula C39H66ClN5O8.
  • Supplied as a solid in milligram quantities for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.