Learn More
Medchemexpress LLC Monomethylauristatin F sodium | 1799706-65-2 | 99.6% | 753.94 g/mol | C39H64N5NaO8 | 100 MG
SDP
Catalog No. 5000204407
Encompass_Preferred
MMAF sodium (Monomethylauristatin F sodium) is the sodium salt of monomethylauristatin F, a potent tubulin polymerization inhibitor used as a cytotoxic payload in antibody-drug conjugates and antitumor research. It is supplied as a white to off-white solid for research use and should be stored refrigerated under inert atmosphere to maintain stability.
  • Potent tubulin polymerization inhibitor.
  • Used as a cytotoxic payload in ADC research.
  • White to off-white solid suitable for analytical and synthetic applications.
  • High purity, approximately 99.6%.
  • Molecular weight 753.94 g/mol; CAS 1799706-65-2.
  • Store refrigerated, protect from light, and keep under inert gas where possible.
Catalog No. 50-002-04407 Supplier Medchemexpress LLC Supplier No. HY15579B100MG
MMAF sodium (Monomethylauristatin F sodium) is the sodium salt of monomethylauristatin F, a potent tubulin polymerization inhibitor used as a cytotoxic payload in antibody-drug conjugates and antitumor research. It is supplied as a white to off-white solid for research use and should be stored refrigerated under inert atmosphere to maintain stability.
  • Potent tubulin polymerization inhibitor.
  • Used as a cytotoxic payload in ADC research.
  • White to off-white solid suitable for analytical and synthetic applications.
  • High purity, approximately 99.6%.
  • Molecular weight 753.94 g/mol; CAS 1799706-65-2.
  • Store refrigerated, protect from light, and keep under inert gas where possible.