Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monomethylauristatin F sodium | 1799706-65-2 | 99.6% | 753.94 g·mol⁻¹ | C39H64N5NaO8 | 5 MG
SDP

Catalog No. 5000204406
Encompass_Preferred

Monomethylauristatin F sodium (MMAF sodium) is the sodium salt of monomethyl auristatin F, a potent tubulin polymerization inhibitor used primarily as a cytotoxic payload in antibody-drug conjugate research and preclinical antitumor studies.

  • Potent tubulin polymerization inhibitor used in cytotoxic payload research.
  • High reported purity (99.6%).
  • Molecular weight 753.94 g·mol⁻¹; formula C39H64N5NaO8.
  • Available in small research quantities such as 5 mg.
  • Store at 4°C, protect from light, and store under inert gas where indicated.
  • Handle as a cytotoxic compound and follow appropriate safety measures and disposal procedures.

Catalog No. 50-002-04406 Supplier Medchemexpress LLC Supplier No. HY15579B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monomethylauristatin F sodium (MMAF sodium) is the sodium salt of monomethyl auristatin F, a potent tubulin polymerization inhibitor used primarily as a cytotoxic payload in antibody-drug conjugate research and preclinical antitumor studies.

  • Potent tubulin polymerization inhibitor used in cytotoxic payload research.
  • High reported purity (99.6%).
  • Molecular weight 753.94 g·mol⁻¹; formula C39H64N5NaO8.
  • Available in small research quantities such as 5 mg.
  • Store at 4°C, protect from light, and store under inert gas where indicated.
  • Handle as a cytotoxic compound and follow appropriate safety measures and disposal procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.