Learn More
Anton Paar Monomode microwave reactor (850 W) with MAS 24 autosampler for up to 300 °C and 30 bar. Compatible with borosilicate or SiC vials. Includes camera, touchscreen, stirring, pressure and IR sensors, plus USB/Ethernet and optional Ruby Thermometer.
Catalog No. 5000580441
Encompass_Preferred
Power supply: 230V, 50 Hz/60 Hz, 1600 VAIncluded:- Autosampler MAS24- Carousel- Consumables Set- Spill bin (83740)- 4 pcs. Reaction Vial G30 Wide Neck- 4 pcs. Snap Cap Wide Neck- 50 pcs. PTFE Septum Wide Neck- 10 pcs. Stir bar oval 15x6 mm- Blind pin (89189)- Adaptor ring (88999)- 2 pcs. Cavity collar (83212)- 3 pcs. Filter disc (84294)- Piston cover key (89196)
Catalog No. 50-005-80441 Supplier Anton Paar Supplier No. 165741
Please contact your sales representative for a quote or further ordering instructions.
Power supply: 230V, 50 Hz/60 Hz, 1600 VAIncluded:- Autosampler MAS24- Carousel- Consumables Set- Spill bin (83740)- 4 pcs. Reaction Vial G30 Wide Neck- 4 pcs. Snap Cap Wide Neck- 50 pcs. PTFE Septum Wide Neck- 10 pcs. Stir bar oval 15x6 mm- Blind pin (89189)- Adaptor ring (88999)- 2 pcs. Cavity collar (83212)- 3 pcs. Filter disc (84294)- Piston cover key (89196)