Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monononadecanoin, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111012486 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2486 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2486 Supplier MilliporeSigma Supelco Supplier No. 0219325MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 112340-30-4
Linear Formula C22H44O4
Synonym Glyceryl monononadecanoate
Type Analytical Standard
Quantity 25 mg
Formula Weight 372.58
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.