Learn More
Promocell GmbH PromoCell Mononuclear Cell Medium 500 ml, ready-to-use, with C-39830 attached to flask
Catalog No. 50306287
Encompass
The PromoCell Mononuclear Cell Medium is a very rich short-term maintenance medium, which provides utmost cell viability for Mononuclear Cells from umbilical cord blood, adult peripheral blood and other sources, e.g. bone marrow.It is suitable to maintain Mononuclear Cells for up to 48 hours before proceeding with your experiments.
Catalog No. 50-306-287 Supplier Promocell GmbH Supplier No. C28030
The PromoCell Mononuclear Cell Medium is a very rich short-term maintenance medium, which provides utmost cell viability for Mononuclear Cells from umbilical cord blood, adult peripheral blood and other sources, e.g. bone marrow.It is suitable to maintain Mononuclear Cells for up to 48 hours before proceeding with your experiments.