Learn More
Medchemexpress LLC Monooctyl phthalate-d4 | 1398065-74-1 | MFCD22565813 | 98.0% | C16H18D4O4 | 10MG
SDP
Catalog No. 5000259202
Encompass_Preferred
Monooctyl phthalate-d4 is a deuterium-labeled internal standard of monooctyl phthalate (CAS 1398065-74-1) intended for quantitative analysis by NMR, GC-MS, and LC-MS. Supplied at high purity in small reference quantities, it is used as a tracer or calibration standard in analytical method development and routine quantitation; for research use only.
  • Deuterium-labeled (d4) internal standard for precise quantitation.
  • High purity (98.0%) suitable for analytical workflows.
  • Compatible with NMR, GC-MS, and LC-MS techniques.
  • Provided in small reference quantities for tracer and calibration use.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-59202 Supplier Medchemexpress LLC Supplier No. HY133671S10MG
Monooctyl phthalate-d4 is a deuterium-labeled internal standard of monooctyl phthalate (CAS 1398065-74-1) intended for quantitative analysis by NMR, GC-MS, and LC-MS. Supplied at high purity in small reference quantities, it is used as a tracer or calibration standard in analytical method development and routine quantitation; for research use only.
  • Deuterium-labeled (d4) internal standard for precise quantitation.
  • High purity (98.0%) suitable for analytical workflows.
  • Compatible with NMR, GC-MS, and LC-MS techniques.
  • Provided in small reference quantities for tracer and calibration use.
  • Intended for research use only; not for human or clinical use.