Learn More
Medchemexpress LLC Monooctyl succinate 100mg | 74295-86-6 | 100 MG
SDP
Catalog No. 5000228199
Encompass_Preferred
Monooctyl succinate is a monoester of succinic acid used as a surfactant and potential fragrance releaser. It is provided for research and formulation studies and is characterized by high purity and defined molecular properties.
  • High purity suitable for research applications.
  • Molecular weight 230.30 g/mol and formula C12H22O4.
  • Available in small pack sizes, including 100 MG.
  • Useful as a formulation component and fragrance-release study material.
Catalog No. 50-002-28199 Supplier Medchemexpress LLC Supplier No. HY114458100MG
Monooctyl succinate is a monoester of succinic acid used as a surfactant and potential fragrance releaser. It is provided for research and formulation studies and is characterized by high purity and defined molecular properties.
  • High purity suitable for research applications.
  • Molecular weight 230.30 g/mol and formula C12H22O4.
  • Available in small pack sizes, including 100 MG.
  • Useful as a formulation component and fragrance-release study material.