Learn More
Medchemexpress LLC Monooctyl succinate 5mg | 74295-86-6 | 5 MG
SDP
Catalog No. 5000228198
Encompass_Preferred
Monooctyl succinate is a monoester used as a surfactant and a potential fragrance releaser for research applications. It is supplied as a high-purity solid for laboratory use.
  • High purity (99.9%) suitable for research applications.
  • Molecular weight 230.30 g/mol and formula C12H22O4.
  • Available in small milligram pack sizes for analytical and formulation work.
  • Commonly used as a surfactant and fragrance-release ingredient in formulation studies.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-28198 Supplier Medchemexpress LLC Supplier No. HY1144585MG
Monooctyl succinate is a monoester used as a surfactant and a potential fragrance releaser for research applications. It is supplied as a high-purity solid for laboratory use.
  • High purity (99.9%) suitable for research applications.
  • Molecular weight 230.30 g/mol and formula C12H22O4.
  • Available in small milligram pack sizes for analytical and formulation work.
  • Commonly used as a surfactant and fragrance-release ingredient in formulation studies.
  • For research use only; not for human or clinical use.