Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monoolein-d7 | 2260669-49-4 | ≥99.0% | 1 MG
SDP

Supplier:  Medchemexpress LLC HY128754S11MG

Encompass_Preferred

Monoolein-d7 is deuterium labeled Monoolein, functioning as an endogenous metabolite. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily serving as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.

  • Deuterium labeled monoolein
  • Functions as an endogenous metabolite
  • Used as tracers for quantitation in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
  • Purity of 99.0% or greater
  • Recommended storage condition: solution at -20°C for up to 2 years

Catalog No. 50-003-39377


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.