Learn More
Avanti Polar Lipids MONOPHOSPHORYL LIPID A 5MG
Catalog No. NC3960505
Encompass
PHAD™-504 was designed as a synthetic structural analog of detoxified MPLA derived from E. coli lipopolysaccharide (LPS). It is structurally similar to PHAD™, differing only in the length of a single fatty acid chain. As expected, the activity of PHAD™-504 is quite similar to that of PHAD™, making the two products interchangeable as adjuvants in vaccine or immunotherapy formulations. purity 99%
Catalog No. NC3960505 Supplier Avanti Polar Lipids Supplier No. 699810P5MG
PHAD™-504 was designed as a synthetic structural analog of detoxified MPLA derived from E. coli lipopolysaccharide (LPS). It is structurally similar to PHAD™, differing only in the length of a single fatty acid chain. As expected, the activity of PHAD™-504 is quite similar to that of PHAD™, making the two products interchangeable as adjuvants in vaccine or immunotherapy formulations. purity 99%