Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Monopropyl phthalate | 4376-19-6 | >98.0% | 208.21 g·mol^-1 | C11H12O4 | 25 MG
SDP

Catalog No. 5000284709
Encompass_Preferred

Monopropyl phthalate is a phthalate ester metabolite formed from hydrolysis of dipropyl phthalate. It is supplied as a research reagent for laboratory analytical use, including metabolic and toxicity studies, and method development.

  • CAS number 4376-19-6 for unambiguous identification.
  • Molecular formula C11H12O4; molecular weight 208.21 g·mol^-1.
  • Available in small, premeasured quantities suitable for analytical standards and reference work.
  • Store under conditions specified in the certificate of analysis to maintain stability.
  • Intended for research use only; not for human or clinical applications.

Catalog No. 50-002-84709 Supplier Medchemexpress LLC Supplier No. HYW70339825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Monopropyl phthalate is a phthalate ester metabolite formed from hydrolysis of dipropyl phthalate. It is supplied as a research reagent for laboratory analytical use, including metabolic and toxicity studies, and method development.

  • CAS number 4376-19-6 for unambiguous identification.
  • Molecular formula C11H12O4; molecular weight 208.21 g·mol^-1.
  • Available in small, premeasured quantities suitable for analytical standards and reference work.
  • Store under conditions specified in the certificate of analysis to maintain stability.
  • Intended for research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.