Learn More
Medchemexpress LLC Monopropyl phthalate | 4376-19-6 | >98.0% | 208.21 g/mol | C11H12O4 | 100 MG
SDP
Catalog No. 5000284676
Encompass_Preferred
Monopropyl phthalate is a phthalate metabolite used as an analytical reference standard for metabolite identification, environmental fate studies, and method development. It is supplied in milligram-scale packages for laboratory use; consult the Certificate of Analysis for batch-specific data and storage instructions.
  • High purity (>98%) suitable for analytical applications.
  • Available in milligram-scale packages (25-500 MG) for standards and small-scale experiments.
  • Molecular formula C11H12O4; molecular weight 208.21 g/mol.
  • Useful for metabolite identification and environmental fate research.
  • Consult the Certificate of Analysis for storage, handling, and batch-specific information.
Catalog No. 50-002-84676 Supplier Medchemexpress LLC Supplier No. HYW703398100MG
Monopropyl phthalate is a phthalate metabolite used as an analytical reference standard for metabolite identification, environmental fate studies, and method development. It is supplied in milligram-scale packages for laboratory use; consult the Certificate of Analysis for batch-specific data and storage instructions.
  • High purity (>98%) suitable for analytical applications.
  • Available in milligram-scale packages (25-500 MG) for standards and small-scale experiments.
  • Molecular formula C11H12O4; molecular weight 208.21 g/mol.
  • Useful for metabolite identification and environmental fate research.
  • Consult the Certificate of Analysis for storage, handling, and batch-specific information.