Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Camelid VHH Antibody [Biotin], mAb
Catalog No. NC1883262
Encompass
GenScript MonoRab Rabbit Anti-Camelid VHH Antibody [Biotin], mAb is specific for Camelid VHH Antibody (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.
Catalog No. NC1883262 Supplier Genscript Corporation Supplier No. A01995200
GenScript MonoRab Rabbit Anti-Camelid VHH Antibody [Biotin], mAb is specific for Camelid VHH Antibody (Alpaca, Llama and Camel). The monoclonal antibody has no cross-reactivity with mouse, rat, rabbit, chicken, goat or human immunoglobulins.