Learn More
Genscript Corporation MonoRab™ Rabbit Anti-Camelid VHH Cocktail
Catalog No. NC1883263
Encompass
GenScript’s MonoRab™ Rabbit Anti-Camelid VHH Cocktail is specific forCamelid VHH antibody (Llama, Alpaca, and Camel). It has no cross-reactivitywith mouse, rat, rabbit, goat or human immunoglobulins. 
Catalog No. NC1883263 Supplier Genscript Corporation Supplier No. A02014200
GenScript’s MonoRab™ Rabbit Anti-Camelid VHH Cocktail is specific forCamelid VHH antibody (Llama, Alpaca, and Camel). It has no cross-reactivitywith mouse, rat, rabbit, goat or human immunoglobulins. 