Learn More
Genscript Corporation Antibody Resin Protein Purification Protein Purification 25 ML 1/EA
Catalog No. 502529986
Encompass
GenScripts MonoRab Anti-Camelid VHH Affinity Resin (Cat. No. L00905) is designed for purification of Camelid VHH single domain antibody from commonly used protein expression systems such as bacteria yeast and mammalian cells. The VHH single domain antibody in cell lysate can specifically bind to the Anti-Camelid VHH monoclonal antibody coupled to the resin. Nonspecific binding reactions are eliminated by stringent washing steps and the purified target protein is eluted with a high recovery rate.
Catalog No. 50-252-9986 Supplier Genscript Corporation Supplier No. L0090525
GenScripts MonoRab Anti-Camelid VHH Affinity Resin (Cat. No. L00905) is designed for purification of Camelid VHH single domain antibody from commonly used protein expression systems such as bacteria yeast and mammalian cells. The VHH single domain antibody in cell lysate can specifically bind to the Anti-Camelid VHH monoclonal antibody coupled to the resin. Nonspecific binding reactions are eliminated by stringent washing steps and the purified target protein is eluted with a high recovery rate.