Learn More
Genscript Corporation MonoRab Anti-Mouse IgG HL
Supplier:  Genscript Corporation V9030110
Encompass
Rabbit anti-mouse monoclonal antibody is a secondary antibody which recognizes mouse immunoglobulins with high affinity It is a useful tool for the detection sorting capture or purification of its specified target It can be used in several analysis systems such as ELISA or Western Blot (WB) Immunohistochemistry (IHC) Flow Cytometry (FACS) and Time-resolved fluorescence immunoassay (TRFIA) It also can be used to develop secondary antibody-coupled magnetic beads for immunoprecipitation (IP) Co-IP and isolation of cells
Catalog No. 50-000-58568