Learn More
Genscript Corporation MonoRab Anti-VHH Affinity Mag
Catalog No. 5000058061
Encompass
This product is specifically designed for purifying camelid and humanized VHH from commonly used protein expression systems such as bacteria yeast and mammalian cells
Catalog No. 50-000-58061 Supplier Genscript Corporation Supplier No. L0103425
This product is specifically designed for purifying camelid and humanized VHH from commonly used protein expression systems such as bacteria yeast and mammalian cells