Learn More
Genscript Corporation MonoRab Anti-VHH Affinity Mag
Supplier:  Genscript Corporation L010341
Encompass
This product is specifically designed for purifying camelid and humanized VHH from commonly used protein expression systems such as bacteria yeast and mammalian cells
Catalog No. 50-000-58397