Learn More
Genscript Corporation MonoRab NT-proBNP 10B11 mA
Catalog No. 5000056797
Encompass
BNP and NT-proBNP are separated from precursor molecule proBNP via proteolytic processing The BNP and NT-proBNP level in blood are proportional to the severity of cardiac dysfunction They can be used for diagnosis of congestive heart failure (CHF)
Catalog No. 50-000-56797 Supplier Genscript Corporation Supplier No. V00705100
Add to Cart
Add to Cart
Request a Quote
BNP and NT-proBNP are separated from precursor molecule proBNP via proteolytic processing The BNP and NT-proBNP level in blood are proportional to the severity of cardiac dysfunction They can be used for diagnosis of congestive heart failure (CHF)